EMAIL THIS PAGE TO A FRIEND
Cell chemical biology
A Fungal-Selective Cytochrome bc
PMID 27524297
Abstract
To cause disease, a microbial pathogen must adapt to the challenges of its host environment. The leading fungal pathogen Candida albicans colonizes nutrient-poor bodily niches, withstands attack from the immune system, and tolerates treatment with azole antifungals, often evolving resistance. To discover agents that block these adaptive strategies, we screened 300,000 compounds for inhibition of azole tolerance in a drug-resistant Candida isolate. We identified a novel indazole derivative that converts azoles from fungistatic to fungicidal drugs by selective inhibition of mitochondrial cytochrome bc
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1850
Inz-1, ≥98% (HPLC)
C16H14N2O2