EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Sensitivity of a tropical micro-crustacean (Daphnia lumholtzi) to trace metals tested in natural water of the Mekong River.
PMID 27528484
Abstract
Metal contamination is one of the major issues to the environment worldwide, yet it is poorly known how exposure to metals affects tropical species. We assessed the sensitivity of a tropical micro-crustacean Daphnia lumholtzi to three trace metals: copper (Cu), zinc (Zn) and nickel (Ni). Both, acute and chronic toxicity tests were conducted with metals dissolved in in situ water collected from two sites in the lower part of the Mekong River. In the acute toxicity test, D. lumholtzi neonates were exposed to Cu (3-30μgL
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36676
Endosulfan sulfate, PESTANAL®, analytical standard
C9H6Cl6O4S