EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Recurrent TRIO Fusion in Nontranslocation-Related Sarcomas.
PMID 27528700
Abstract
Despite various differences, nontranslocation-related sarcomas (e.g., comprising undifferentiated pleomorphic sarcoma, leiomyosarcoma, myxofibrosarcoma) are unified by their complex genetics. Extensive analysis of the tumor genome using molecular cytogenetic approaches showed many chromosomal gains, losses, and translocations per cell. Genomic quantitative alterations and expression variations have been extensively studied by adapted high-throughput approaches, yet translocations still remained unscreened. We therefore analyzed 117 nontranslocation-related sarcomas by RNA sequencing to identify fusion genes. We performed RNA sequencing and applied a bioinformatics pipeline dedicated to the detection of fusion transcripts. RT-PCR and Sanger sequencing were then applied to validate predictions and to search for recurrence and specificity. Among the 6,772 predicted fusion genes, 420 were in-frame. One recurrent rearrangement, consistently involving TRIO with various partners, was identified in 5.1% of cases. TRIO translocations are either intrachromosomal with TERT or interchromosomal with LINC01504 or ZNF558 Our results suggest that all translocations led to a truncated TRIO protein either directly or indirectly by alternative splicing. TRIO rearrangement is associated with a modified transcriptomic program to immunity/inflammation, proliferation and migration, and an increase in proliferation. TRIO fusions have been identified in four different sarcoma histotypes, likely meaning that they are not related to a primary oncogenic event but rather to a secondary one implicated in tumor progression. Moreover, they appear to be specific to nontranslocation-related sarcomas, as no such rearrangement was identified in sarcomas with simple genetics. More cases could lead to a significant association of these fusions to a specific clinical behavior. Clin Cancer Res; 23(3); 857-67. ©2016 AACR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3072 Doxycycline Hydrochloride, Ready Made Solution
C22H24N2O8 · HCl