EMAIL THIS PAGE TO A FRIEND
Nature communications
Asymmetric counteranion-directed Lewis acid organocatalysis for the scalable cyanosilylation of aldehydes.
PMID 27530470
Abstract
Due to the high versatility of chiral cyanohydrins, the catalytic asymmetric cyanation reaction of carbonyl compounds has attracted widespread interest. However, efficient protocols that function at a preparative scale with low catalyst loading are still rare. Here, asymmetric counteranion-directed Lewis acid organocatalysis proves to be remarkably successful in addressing this problem and enabled a molar-scale cyanosilylation in quantitative yield and with excellent enantioselectivity. Also, the catalyst loading could be lowered to a part-per-million level (50 ppm: 0.005 mol%). A readily accessible chiral disulfonimide was used, which in combination with trimethylsilyl cyanide, turned into the active silylium Lewis acid organocatalyst. The nature of a peculiar phenomenon referred to as a "dormant period", which is mainly induced by water, was systematically investigated by means of in situ Fourier transform infrared analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N206
2-Naphthaldehyde, 98%
C11H8O