EMAIL THIS PAGE TO A FRIEND
Journal of the American Society for Mass Spectrometry
Thermodynamic Analysis of the Geldanamycin-Hsp90 Interaction in a Whole Cell Lysate Using a Mass Spectrometry-Based Proteomics Approach.
PMID 27530778
Abstract
Geldanamycin is a natural product with well-established and potent anti-cancer activities. Heat shock protein 90 (Hsp90) is the known target of geldanamycin, which directly binds to Hsp90's N-terminal ATP binding domain and inhibits Hsp90's ATPase activity. The affinity of geldanamycin for Hsp90 has been measured in multiple studies. However, there have been large discrepancies between the reported dissociation constants (i.e., Kd values), which have ranged from low nanomolar to micromolar. Here the stability of proteins from rates of oxidation (SPROX) technique was used in combination with an isobaric mass tagging strategy to measure the binding affinity of geldanamycin to unpurified Hsp90 in an MCF-7 cell lysate. The Kd values determined here were dependent on how long geldanamycin was equilibrated with the lysate prior to SPROX analysis. The Kd values determined using equilibration times of 0.5 and 24 h were 1 and 0.03 μM, respectively. These Kd values, which are similar to those previously reported in a geldanamycin-Hsp90 binding study that involved the use of a fluorescently labeled geldanamycin analogue, establish that the slow-tight binding behavior previously observed for the fluorescently labeled geldanamycin analogue is not an artifact of the fluorescent label, but rather an inherent property of the geldanamycin-Hsp90 binding interaction. The slow-tight binding property of this complex may be related to time-dependent conformational changes in Hsp90 and/or to time-dependent chemical changes in geldanamycin, both of which have been previously proposed to explain the slow-tight binding behavior of the geldanamycin-Hsp90 complex. Graphical Abstract ᅟ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86600
Tetraethylammonium borohydride, technical, ≥95% (T)
C8H24BN