EMAIL THIS PAGE TO A FRIEND
The Journal of physiology
Mechanical activation of angiotensin II type 1 receptors causes actin remodelling and myogenic responsiveness in skeletal muscle arterioles.
PMID 27531064
Abstract
Candesartan, an inverse agonist of the type 1 angiotensin II receptor (AT The G