EMAIL THIS PAGE TO A FRIEND
Journal of neurovirology
Development and characterization of a human microglia cell model of HIV-1 infection.
PMID 27538994
Abstract
Microglia cells are the major reservoir of HIV-1 (HIV) within the CNS. However, current models using transformed cell lines are not representative of primary microglia and fetal brain samples for isolation of primary human microglia (HMG) are increasingly difficult to obtain. Here, we describe a monocyte-derived microglia (MMG) cell model of HIV infection that recapitulates infection of primary HMG. CD14