Environmental toxicology
Exposure to PM2.5 causes genetic changes in fetal rat cerebral cortex and hippocampus.
PMID 27539004
PM2.5 travels along the respiratory tract and enters systemic blood circulation. Studies have shown that PM2.5 increases the incidence of various diseases not only in adults but also in newborn infants. It causes chronic inflammation in pregnant women and retards fetal development. In this study, pregnant rats were exposed to PM2.5 for extended periods of time and it was found that PM2.5 exposure increased immune cells in mother rats. In addition, cytokines and free radicals rapidly accumulated in the amniotic fluid and indirectly affected the fetuses. The authors collected cerebral cortex and hippocampus samples at E18 and analyzed changes of miRNA levels. Expression levels of cortical miR-6315, miR-3588, and miR-466b-5p were upregulated, and positively correlated with the genes Pkn2 (astrocyte migration), Gorab (neuritogenesis), and Mobp (allergic encephalomyelitis). In contrast, PM2.5 decreased expression of miR-338-5p and let-7e-5p, both related to mental development. Further, PM2.5 exposure increased miR-3560 and let-7b-5p in the hippocampus, two proteins that regulate genes Oxct1 and Lin28b that control ketogenesis and glycosylation, and neural cell differentiation, respectively. miR-99b-5p, miR-92b-5p, and miR-99a-5p were decreased, leading to reduced expression of Kbtbd8 and Adam11 which reduced cell mitosis, migration, and differentiation, and inhibited learning abilities and motor coordination of the fetus. © 2016 Wiley Periodicals, Inc. Environ Toxicol 32: 1412-1425, 2017.
Related Materials
Product #
Molecular Formula
Add to Cart
CRM558 Diesel - Clay Loam 1, certified reference material, pkg of 100 g