EMAIL THIS PAGE TO A FRIEND
International journal of radiation biology
Effects of IL-4 on pulmonary fibrosis and the accumulation and phenotype of macrophage subpopulations following thoracic irradiation.
PMID 27539247
Abstract
Thoracic irradiation injures lung parenchyma, triggering inflammation and immune cell activation, leading to pneumonitis and fibrosis. Macrophage polarization contributes to these processes. Since IL-4 promotes pro-fibrotic macrophage activation, its role in radiation-induced lung injury was investigated. Lung macrophage subpopulations were characterized from 3-26 weeks following exposure of WT and IL-4-/- mice to 0 or 12.5 Gray single dose thoracic irradiation. Loss of IL-4 did not prevent fibrosis, but blunted macrophage accumulation within the parenchyma. At 3 weeks following exposure, cell numbers and expression of F4/80 and CD206, an alternative activation marker, decreased in alveolar macrophages but increased in infiltrating macrophages in WT mice. Loss of IL-4 impaired recovery of these markers in alveolar macrophages and blunted expansion of these populations in infiltrating macrophages. CD206+ cells were evident in fibrotic regions of WT mice only, however Arg-1+ cells increased in fibrotic regions in IL-4-/- mice only. Radiation-induced proinflammatory Ly6C expression was more apparent in alveolar and interstitial macrophages from IL-4-/- mice. IL-4 loss did not prevent alternative macrophage activation and fibrosis in irradiated mice. Instead, a role is indicated for IL-4 in maintenance of macrophage populations in the lung following high single dose thoracic irradiation.