EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Quality control slide for potassium hydroxide and cellufluor fungal preparations.
PMID 2754012
Abstract
An opaque, water-insoluble quality control material with a skinlike microscopic appearance was prepared by inoculating melted xanthine (0.4%) agar with filamentous fungi and dispensing drops onto glass slides. After solidification of the agar, the material was rapidly cleared by 10% KOH, revealing fungal elements stained by Cellufluor reagent.