EMAIL THIS PAGE TO A FRIEND
Chemical biology & drug design
Synthesis and biological evaluation of novel flavanone derivatives as potential antipsychotic agents.
PMID 27543496
Abstract
In this study, a series of novel flavanone derivatives were designed and synthesized, and the antipsychotic activities of the target compounds were evaluated in vitro and in vivo. The results showed that synthesized compounds 7a-7g decreased the activity of dopamine D
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3262
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
C5H9NO4