EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Inhibitory Interaction Potential of 22 Antituberculosis Drugs on Organic Anion and Cation Transporters of the SLC22A Family.
PMID 27550354
Abstract
Twenty-two currently marketed antituberculosis drugs were comprehensively evaluated for their inhibitory effect on organic anionic transporter (OAT)- and organic cation transporter (OCT)-mediated uptake using stably transfected HEK293 cells in vitro We observed moderate to strong inhibitory effects on OAT1- and OAT3-mediated para-aminohippurate (PAH) uptake and OCT1- and OCT2-mediated N-methyl-4-phenylpylidinium acetate (MPP
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0533
Rifapentine
C47H64N4O12