EMAIL THIS PAGE TO A FRIEND
Journal of analytical toxicology
Quantification of selected herbicides and chlorinated phenols in urine by using gas chromatography/mass spectrometry/mass spectrometry.
PMID 2755087
Abstract
We have developed a method for determining selected chlorinated phenols and phenoxy herbicides in urine. The process of preparing the samples includes acid hydrolysis, extraction with benzene, derivatization with diazoethane, and column chromatography cleanup. We quantify the more volatile compounds by using capillary column gas chromatography/positive chemical ionization/mass spectrometry/mass spectrometry. Less volatile compounds are quantified by using electron capture negative chemical ionization in a single stage mass spectrometry mode. Quality control samples are included in each analytical run, and the results demonstrate that the analytical system is in control. Positive values for the target analytes are determined on the basis of appropriate relative retention time, a signal-to-noise ratio greater than 3:1, and a calculated concentration greater than 1 ppb. We determine the chlorine isotope ratios for each compound to assess the presence or absence of interferences. This analytical method has been applied in a case-control study of 199 individuals to examine exposure to the 12 target analytes.