EMAIL THIS PAGE TO A FRIEND
Molecular cancer therapeutics
TLR4-Dependent Claudin-1 Internalization and Secretagogue-Mediated Chloride Secretion Regulate Irinotecan-Induced Diarrhea.
PMID 27550942
Abstract
We have previously shown increased intestinal permeability, to 4-kDa FITC-dextran, in BALB/c mice treated with irinotecan. Importantly, genetic deletion of Toll-like receptor 4 (TLR4; Tlr4