EMAIL THIS PAGE TO A FRIEND
The journal of physiological sciences : JPS
Elevated C-type natriuretic peptide elicits exercise preconditioning-induced cardioprotection against myocardial injury probably via the up-regulation of NPR-B.
PMID 27557795
Abstract
To evaluate exercise preconditioning (EP)-induced cardioprotective effects against exercise-induced acute myocardial injury and investigate the alterations of C-type natriuretic peptide (CNP) and its specific receptor, natriuretic peptide receptor B (NPR-B), during EP-induced cardioprotection. Rats were subjected to treadmill exercise as an EP model (4 periods of 10 min each at 30 m/min with intervening periods of rest lasting 10 min). High-intensity exercise was performed 0.5 and 24 h after the EP. EP attenuated high-intensity exercise-induced myocardial injury in both the early and late phases. After EP and high-intensity exercise, CNP and NPR-B levels increased robustly, but no alterations in the plasma CNP were observed. The enhanced NPR-B, plasma and tissue CNP, and its mRNA levels after high-intensity exercise were significantly elevated by EP. These results suggest that cardiac CNP and NPR-B play an important role in EP-mediated cardioprotection against high-intensity exercise-induced myocardial injury in rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8768
Natriuretic Peptide, C-Type, ≥95% (HPLC)
C93H157N27O28S3