EMAIL THIS PAGE TO A FRIEND
Current topics in medicinal chemistry
Elucidation of Phosphodiesterase-1 Inhibitory Effect of Some Selected Natural Polyphenolics Using In Vitro and In Silico Methods.
PMID 27558680
Abstract
Polyphenols are organic compounds widely found in plants and possess many diverse biological activities. In the current study, eleven polyphenolic compounds [rutin (1), trans-ferulic acid (2), epigallaocatechin gallate (3), fumaric acid (4), chlorogenic acid (5), gallic acid (6), trans-resveratrol (7), propyl gallate (8), (+)-catechin (9), quercetin (10), and kaempferol-3-O-β-D-galactoside (11)] have been evaluated for their in vitro inhibitory activity against phosphodiesterase- 1 (PDE-1). The active compounds 1, 5, 6, and 7 possessed IC50 values of 173.90 ± 2.58, 36.67 ± 1.60, 325.19 ± 3.62, and 323.81 ± 4.25 μM, respectively. In silico experiments were performed to elucidate the binding patterns of inhibitor molecules with the active sites of the enzymes. Molecular docking studies have shown that compounds 1 and 5 have better docking results than standard inhibitor (EDTA) against the PDE-1. In conclusion, chlorogenic acid, rutin, gallic acid, and resveratrol could be promising PDE-1 inhibitors and leads to identify new drug candidates against PDE-1-associated disorders.
Related Materials