EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Characterization of the effects of endotoxin on macrophage tumor cell killing.
PMID 27559
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5399
Lipid A, diphosphoryl from Escherichia coli F583 (Rd mutant)