EMAIL THIS PAGE TO A FRIEND
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Composite metric R
PMID 27563836
Abstract
This study looked to investigate a new quantitative metric, R