EMAIL THIS PAGE TO A FRIEND
Biological trace element research
A Selenium-Enriched Ziyang Green Tea Polysaccharide Induces Bax-Dependent Mitochondrial Apoptosis and Inhibits TGF-β1-Stimulated Collagen Expression in Human Keloid Fibroblasts via NG2 Inactivation.
PMID 27565798
Abstract
Keloids are fibroproliferative disorders characterized by the overabundant deposition of extracellular matrix (ECM), especially collagen and overgrowth of scar tissue in response to cutaneous injury. In this study, we isolated a selenium (Se)-containing polysaccharide (Se-ZGTP-I) from Ziyang green tea and explored its potential therapeutic effects on keloid fibroblasts formation. 3-(4,5-Dimethyl-thiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) and annexin V/propidium iodide (PI) staining assays demonstrated that Se-ZGTP-I or neuron-glia 2 (NG2) short hairpin RNA (shRNA) significantly inhibited proliferation of human keloid fibroblasts via induction of apoptosis. Besides, the activation of caspase-3 and the subsequent cleavage of poly (ADP-ribose) polymerase (PARP) were observed in keloid fibroblasts following Se-ZGTP-I (200 and 400 μg/ml) or NG2 shRNA treatment. Moreover, Western blotting analysis showed that treatment of keloid fibroblasts with Se-ZGTP-I (200 and 400 μg/ml) or NG2 shRNA resulted in an increase of pro-apoptotic protein Bax expression and a decrease in expression levels of anti-apoptotic protein Bcl-2 and NG2. In addition, type I collagen biosynthesis and protein expression in keloid fibroblasts following TGF-β1 stimulation were decreased by Se-ZGTP-I (200 and 400 μg/ml) or NG2 shRNA management. Current findings imply that Se-ZGTP-I has a therapeutic potential to intervene and prevent keloid formation and other fibrotic diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W373011 L-Rhamnose, natural sourced, 99%, FG
C6H12O5H2O