EMAIL THIS PAGE TO A FRIEND
Brain research bulletin
Postnatal changes in glutamatergic inputs of jaw-closing motoneuron dendrites.
PMID 27568204
Abstract
Dendrites of masseter (jaw-closing) motoneurons (MMNs) are well developed and ramify extensively throughout the trigeminal motor nucleus and often extend into the adjacent reticular formation. It is possible that the dendrites have active properties, which are altered with the development of the orofacial musculoskeletal system. Thus, we examined the changes in somatic voltage responses evoked by photostimulation of the MMN dendrites by laser photolysis of caged glutamate from postnatal day (P) 2-5 and 9-12 rats. We photostimulated 39 spots that were arranged around each recorded neuron in a concave shape and found that the dendritic stimulation induced somatic depolarization in the presence of tetrodotoxin in all MMNs. With increasing photostimulation intensity, the responses grew in amplitude up to a certain threshold, where a step-like increase in amplitude occurred. In 75% of P2-5 MMNs, the step-like increase in amplitude, which was blocked by 20μM D(-)-2-amino-5-phosphonovaleric acid application, corresponded to the NMDA spikes/plateau potentials. In contrast, at P9-12 the responses became significantly smaller in amplitude and shorter in duration and only one neuron out of 12 MMNs showed NMDA spikes/plateau potentials. These results suggest that the glutamatergic responses evoked by photostimulation of the MMN dendrites change during the first two postnatal weeks, and these changes may be involved in the transition from suckling to chewing during postnatal development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4261
Biocytin, ≥98% (TLC)
C16H28N4O4S
M3262
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
C5H9NO4