EMAIL THIS PAGE TO A FRIEND
Leukemia
The immune receptor Trem1 cooperates with diminished DNA damage response to induce preleukemic stem cell expansion.
PMID 27568523
Abstract
Fanconi anemia (FA) is an inherited bone marrow failure syndrome with extremely high risk of leukemic transformation. Here we investigate the relationship between DNA damage response (DDR) and leukemogenesis using the Fanca knockout mouse model. We found that chronic exposure of the Fanca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
248401
Magnesium methyl carbonate solution, 2.0 M in DMF
C3H6MgO4