EMAIL THIS PAGE TO A FRIEND
Glia
TRPA1 deficiency is protective in cuprizone-induced demyelination-A new target against oligodendrocyte apoptosis.
PMID 27568827
Abstract
Multiple sclerosis is a chronic inflammatory, demyelinating degenerative disease of the central nervous system. Current treatments target pathological immune responses to counteract the inflammatory processes. However, these drugs do not restrain the long-term progression of clinical disability. For this reason, new therapeutic approaches and identification of novel target molecules are needed to prevent demyelination or promote repair mechanisms. Transient Receptor Potential Ankyrin 1 (TRPA1) is a nonselective cation channel with relatively high Ca