EMAIL THIS PAGE TO A FRIEND
Molecular endocrinology (Baltimore, Md.)
Phosphorylation of Farnesoid X Receptor at Serine 154 Links Ligand Activation With Degradation.
PMID 27571290
Abstract
Comparison of 11 human nuclear receptor amino acid sequences revealed a conserved phosphorylation motif within their DNA-binding domains as an intramolecular signal that regulates proteolytic degradation. Nuclear receptors use this signal to either degrade or proscribe degradation through either the proteasome or nonproteasome pathways. A phosphomimetic farnesoid X receptor (FXR) S154D mutant neither bound to nor trans-activated an FXR-response element-driven reporter gene and was rapidly degraded in COS-1 cells. Ectopically expressed FXR had increased Ser154 phosphorylation in COS-1 cells after ligand treatment, and knock-down of the nuclear vaccinia-related kinase 1 (VRK1) greatly reduced this phosphorylation. FXR was phosphorylated at Ser154 in the nucleus of centrilobular hepatocytes only in ligand-treated mice. Thus, FXR Ser154 phosphorylation is a rheostat for activation and subsequent degradation that controls receptor levels and activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5172
GW4064, ≥97% (HPLC)
C28H22Cl3NO4
EHU119541 MISSION® esiRNA, esiRNA human VRK1 (esiRNA1)