EMAIL THIS PAGE TO A FRIEND
The Analyst
Extractive spectrophotometric determination of some anti-inflammatory agents with methylene violet.
PMID 2757224
Abstract
A fairly sensitive spectrophotometric method for the determination of ibuprofen, ketoprofen, piroxicam, diclofenac sodium, mefenamic acid or enfenamic acid in bulk samples and pharmaceutical preparations is described, based on the formation of a chloroform-soluble, coloured ion-association complex between the drug and Methylene Violet at pH 7.6.
Related Materials