EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
High-Sulfur-Vacancy Amorphous Molybdenum Sulfide as a High Current Electrocatalyst in Hydrogen Evolution.
PMID 27578319
Abstract
The remote hydrogen plasma is able to create abundant S-vacancies on amorphous molybdenum sulfide (a-MoS