EMAIL THIS PAGE TO A FRIEND
Cell
Two structural genes on different chromosomes are required for encoding the major subunit of human red cell glucose-6-phosphate dehydrogenase.
PMID 2758468
Abstract
Structural analysis revealed the existence of two types of subunits in human red cell glucose-6-phosphate dehydrogenase. The two subunits have the same COOH region consisting of 479 amino acid residues, but their NH2-terminal regions are different in size and sequence. The minor subunit can be fully encoded by the X-linked G6PD cDNA, but the NH2-terminal region of the major subunit cannot. The cDNA and the gene for the NH2-terminal region of the major subunit were cloned and characterized. Southern blot hybridization indicated that the gene for the NH2-terminal region is on chromosome 6, not on the X chromosome. Northern blot hybridization demonstrated an existence of two separate mRNA components, one for the COOH-terminal region and the other for the NH2-terminal region. Two separate structural genes, the X-linked and chromosome 6-linked genes, must be coresponsible for encoding the single chain subunit. Either cross-translation of two mRNAs, or transpeptidation, or some other mechanism must be involved in the synthesis of human red cell G6PD.