EMAIL THIS PAGE TO A FRIEND
La Radiologia medica
Usefulness of paclitaxel-releasing high-pressure balloon associated with cutting balloon angioplasty for treatment of outflow stenoses of failing hemodialysis arteriovenous shunts.
PMID 27601144
Abstract
To evaluate the technical and clinical success, primary patency (PP) and complications of angioplasty performed with paclitaxel-coated balloon (PCBs) associated with cutting balloon and for the treatment of the outflow stenoses of failing hemodialysis arteriovenous shunt. From September 2014 to September 2015, 50 patients with 66 stenoses were registered. Vascular accesses were autogenous (nxa0=xa020) and prosthetic (nxa0=xa030). Stenosis were documented during follow-up with routine echo-color Doppler, clinical evaluation and in the remaining incidentally during fistulography. Angioplasty was performed with cutting balloon and afterward with PCB. The mean follow-up time was 8xa0months (range 6-15xa0months). Technical success, clinical success, primary patency and complications were registered. Technical success was 100xa0%. Clinical success was 94.7xa0%. Primary patency rate was 87.7xa0%; in five patients, a significant re-stenosis (≥50xa0%) was registered. A residual asymptomatic stenosis (<30xa0%) was registered in four cases (7xa0%). No major complications were registered. A short-term patency benefit may be obtained including PCB in angioplasty treatment of failing hemodialysis arteriovenous shunts.