EMAIL THIS PAGE TO A FRIEND
Magnetic resonance in medicine
Application of dual
PMID 27610596
Abstract
Cellular MRI) was used to detect implanted human mesenchymal stem cells (hMSCs) and the resulting macrophage infiltration that occurs in response to xenotransplantation. Human mesenchymal stem cells were prelabeled with a fluorine-19 ( Quantification of A dual-labeling technique was used to noninvasively track two distinct cell populations simultaneously. This information could be used to provide additional insight into the cause of graft failure. Magn Reson Med 78:713-720, 2017. © 2016 International Society for Magnetic Resonance in Medicine.