EMAIL THIS PAGE TO A FRIEND
Cell proliferation
p53-dependent up-regulation of CDKN1A and down-regulation of CCNE2 in response to beryllium.
PMID 27611480
Abstract
Beryllium salts (here, beryllium sulphate) can produce a cytostatic effect in some cell types. The basis for this effect may include increased expression of proliferation inhibitors, reduced expression of proliferation promoters, or both. This study sought to determine the role of p53, the tumour-suppressing transcription factor, in mediating beryllium-induced cytostasis. Human A172 glioma cells express wild-type TP53 gene. Activity of p53 was experimentally manipulated using siRNA and related approaches. Key elements of the beryllium-response were compared in normal and p53-knockdown A172 cells using RT-PCR and Western blotting. In A172 cells, 10 μm BeSO Beryllium elicited p53-dependent changes in mRNA levels of key determinants of cell proliferation such as p21 and cyclin E2. However, cyclin E2 protein appeared to be aberrantly regulated in this cell type, as its turnover was unexpectedly slow.