EMAIL THIS PAGE TO A FRIEND
Stem cells and development
Sodium Tungstate for Promoting Mesenchymal Stem Cell Chondrogenesis.
PMID 27615276
Abstract
Articular cartilage has a limited ability to heal. Mesenchymal stem cells (MSCs) derived from the bone marrow have shown promise as a cell type for cartilage regeneration strategies. In this study, sodium tungstate (Na