EMAIL THIS PAGE TO A FRIEND
Fish physiology and biochemistry
Characterization of the isoforms of type IIb sodium-dependent phosphate cotransporter (Slc34a2) in yellow catfish, Pelteobagrus fulvidraco, and their vitamin D
PMID 27620186
Abstract
In this study, two isoforms slc34a2 genes (type IIb sodium-dependent phosphate cotransporter), slc34a2a2 and slc34a2b, were cloned from intestine and kidney of yellow catfish (Pelteobagrus fulvidraco), with rapid amplification of cDNA ends. The structure differences and the regulation effects of dietary VD
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5669
Pyridoxine, ≥98%
C8H11NO3