EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Cytotoxic ethnic Yao medicine Baizuan, leaves of Schisandra viridis A. C. Smith.
PMID 27620660
Abstract
The ethnic Chinese Yao medicine Baizuan, which are the leaves of Schisandra viridis A. C. Smith, is traditionally used, in combination with other herbs, to soften hard lumps and dispel nodes in the treatment of cancer, however, this property has not been well studied with a clear indication of the active principles. The experiments were carried out to investigate the cytotoxic activity of the extracts and to identify the active principles from the extract, which could support the traditional application of treating cancer. Dried and ground plant material was extracted with water and ethanol and further purified by HPLC. The cytotoxicity of the extracts, fractions and pure compounds were evaluated for their abilities to inhibit the proliferation of breast cancer cells MCF7 and tongue cancer cells CAL27. The cytotoxicity of the pure compounds were also tested against Human Embryonic Kidney cell line HEK293. Both aqueous and ethanol extracts showed activities against MCF7 and CAL27 cancer cells. Bioassay-guided fractionation and purification of the extracts resulted in six active principles, including five dibenzocyclooctene lignans namely gomisin H (1), schisandrin (2), angeloylgomisin H (3), (+)-gomisin M The traditional extraction protocol using boiled water afforded three moderately active lignans 1-3. Ethanol extraction, which is widely used in the preparation of herbal remedies in China, yielded three additional active compounds 4-5 with more potent activities. These results provided a rationale for the traditional application of the ethnic Yao medicine Baizuan in the treatment of cancer.