EMAIL THIS PAGE TO A FRIEND
Traffic (Copenhagen, Denmark)
Cresyl violet: a superior fluorescent lysosomal marker.
PMID 27621028
Abstract
We have characterized cresyl violet as a membrane-permeant fluorophore that localizes to lysosomes and acidic vacuoles of budding yeast, Drosophila, human, murine and canine cells. An acidotropic weak base, cresyl violet is shown to be virtually insensitive to physiological alkali and divalent cations. Because of its unique spectral properties, it can be used in combination with green, red and far-red fluorophores, is less susceptible to photobleaching than alternative acidotropic probes, and does not undergo photoconversion. At concentrations that yield bright labeling of acidic compartments, cresyl violet does not alter the organellar pH nor does it affect the buffering capacity. Its affordability, together with its chemical and spectral properties, make cresyl violet a superior lysosomal marker devoid of many of the negative characteristics associated with other lysosomal probes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S6814
SynaptoGreen C4, ≥95% (HPLC), solid
C30H49Br2N3