EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Ultrasound-Assisted Surfactant-Enhanced Emulsification Micro-Extraction Followed by HPLC for Determination of Preservatives in Water, Beverages and Personal Care Products.
PMID 27621134
Abstract
An ultrasound-assisted surfactant-enhanced emulsification micro-extraction (UASEME) procedure has been developed for pre-concentration of benzoic acid (BA) and paraben preservatives, including methylparaben, ethylparaben, propylparaben and butylparaben, prior to high-performance liquid chromatography-ultraviolet (HPLC-UV) analysis. Separations were performed on a Lichrospher RP-18 endcapped 5 µm, using an isocratic mobile phase of 40% acetonitile, at a flow rate of 1 mL min