EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Oleoylethanolamine and palmitoylethanolamine modulate intestinal permeability in vitro via TRPV1 and PPARα.
PMID 27623929
Abstract
Cannabinoids modulate intestinal permeability through cannabinoid receptor 1 (CB