EMAIL THIS PAGE TO A FRIEND
Planta medica
Curcumin Encapsulated into Methoxy Poly(Ethylene Glycol) Poly(ε-Caprolactone) Nanoparticles Increases Cellular Uptake and Neuroprotective Effect in Glioma Cells.
PMID 27626946
Abstract
Curcumin is a natural polyphenolic compound isolated from turmeric (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
704067
ε-Caprolactone, 97%
C6H10O2
Q4951
Quercetin, ≥95% (HPLC), solid
C15H10O7