EMAIL THIS PAGE TO A FRIEND
Bulletin of entomological research
Chemical investigations of volatile kairomones produced by Hyphantria cunea (Drury), a host of the parasitoid Chouioia cunea Yang.
PMID 27628497
Abstract
In tritrophic 'plants-herbivores-natural enemies' systems, there are relatively few reports concerning the role(s) of kairomones in pupal parasitism. Chouioia cunea Yang (Hymenoptera: Eulophidae), an endoparasitic chalcid wasp, parasitizes pupae of the fall webworm (Hyphantria cunea Drury). The role of host-related kairomones was investigated using electroantennogram (EAG) and behavioral techniques. Chemicals from some host stages (pupae) and host by-products (frass), induced arrestment behavior of female parasitoids, while chemicals from prepupae, were inactive. Gas chromatography-mass spectrometry analysis of volatiles collected from pupae, frass and prepupae using solid-phase microextration revealed seven compounds with carbon chain lengths ranging from C4 to C20. All of the chemicals elicited significant EAG responses in C. cunea. Y-tube olfactometer bioassays demonstrated a significant positive response of mated female C. cunea to 1-dodecene. These data provide a better understanding of the host location mechanisms of pupal parasitoid.