EMAIL THIS PAGE TO A FRIEND
Internal medicine (Tokyo, Japan)
Membranous Nephropathy with an Enhanced Granular Expression of Thrombospondin Type-1 Domain-containing 7A in a Pregnant Woman.
PMID 27629964
Abstract
A 30-year-old woman with proteinuria first noted at 26 weeks of gestation was admitted to undergo further evaluation. A renal biopsy revealed membranous nephropathy (MN). There was no evidence of any secondary MN. Prednisolone was initiated 6 months after delivery. Four months later, her urine protein became negative. Enhanced granular staining for thrombospondin type-1 domain-containing 7A (THSD7A) in the glomeruli was retrospectively detected in a biopsy specimen. A literature review revealed that 60% of cases of THSD7A-related MN occurred in women of childbearing age. Therefore, THSD7A-related MN should be considered in female patients presenting with idiopathic MN in childbearing age.