EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Chiral recognition of penicillamine enantiomers using hemoglobin and gold nanoparticles functionalized graphite-like carbon nitride nanosheets via electrochemiluminescence.
PMID 27632698
Abstract
A new stable and stereo-selective electrochemiluminescence (ECL) interface has been designed for specific recognition of penicillamine (Pen) enantiomers by using hemoglobin (Hb) and gold nanoparticles functionalized graphite-like carbon nitride nanosheets composite (Au-g-C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196312
L-Penicillamine, 99%
C5H11NO2S