EMAIL THIS PAGE TO A FRIEND
The Journal of physiology
Impaired central respiratory chemoreflex in an experimental genetic model of epilepsy.
PMID 27633663
Abstract
It is recognized that seizures commonly cause apnoea and oxygen desaturation, but there is still a lack in the literature about the respiratory impairments observed ictally and in the post-ictal period. Respiratory disorders may involve changes in serotonergic transmission at the level of the retrotrapezoid nucleus (RTN). In this study, we evaluated breathing activity and the role of serotonergic transmission in the RTN with a rat model of tonic-clonic seizures, the Wistar audiogenic rat (WAR). We conclude that the respiratory impairment in the WAR could be correlated to an overall decrease in the number of neurons located in the respiratory column. Respiratory disorders may involve changes in serotonergic neurotransmission at the level of the chemosensitive neurons located in the retrotrapezoid nucleus (RTN). Here, we investigated the central respiratory chemoreflex and the role of serotonergic neurotransmission in the RTN with a rat model of tonic-clonic seizures, the Wistar audiogenic rat (WAR). We found that naive or kindled WARs have reduced resting ventilation and ventilatory response to hypercapnia (7% CO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2886
ExtrAvidin®−Peroxidase, buffered aqueous solution