Journal of cancer research and clinical oncology
Reduced expression of CXCR4, a novel renal cancer stem cell marker, is associated with high-grade renal cell carcinoma.
PMID 27638770
Cancer stem cells (CSCs) represent a population with tumour-initiating, self-renewal, and differentiation potential. This study aimed to evaluate the expression patterns and clinical significance of chemokine receptor type 4 (CXCR4) as a novel CSC marker in renal cell carcinoma (RCC). The expression of CXCR4 was examined in 173 well-defined renal tumour tissues, including 106 (61.5xa0%) clear cell renal cell carcinomas (ccRCCs), 35 (20xa0%) papillary renal cell carcinomas (pRCCs), and 32 (18.5xa0%) chromophobe renal cell carcinomas (ChRCCs), by immunohistochemistry on a tissue microarray. The association between expression of this marker and clinicopathologic parameters was then analysed. There was a significant difference in the expression levels of CXCR4 in the ccRCC samples compared to the ChRCC and pRCC samples (Pxa0