EMAIL THIS PAGE TO A FRIEND
Biotechnology and bioengineering
Metabolic profile of 1,5-diaminopentane producing Corynebacterium glutamicum under scale-down conditions: Blueprint for robustness to bioreactor inhomogeneities.
PMID 27641904
Abstract
Performance losses during scale-up are described since decades, but are still one of the major obstacles for industrial bioprocess development. Consequently, robustness to inhomogeneous cultivation environments is an important quality of industrial production organisms. Especially, Corynebacterium glutamicum was proven to have an outstanding resistance against rapid changes of oxygen and substrate availability as occurring in industrial scale bioreactors. This study focuses on the identification of metabolic key mechanisms for this robustness to get a deeper insight and provide future targets for process orientated strain development. A 1,5-diaminopentane producing C. glutamicum strain was cultivated in a two compartment scale-down device to create short-term environmental changes simulating industrial scale cultivation conditions. Using multi omics based methods, it is shown, that central metabolism is flexibly rearranged under short-term oxygen depletion and carbon source excess to overcome shortage in NAD
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
320684
3,3′-Diethyloxacarbocyanine iodide, 98%
C21H21IN2O2