EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Promotion of osteointegration under diabetic conditions by tantalum coating-based surface modification on 3-dimensional printed porous titanium implants.
PMID 27648775
Abstract
Clinical evidence indicates a high failure rate for titanium implants (TiI) in diabetic patients, involving the overproduction of reactive oxygen species (ROS) at the implant/bone interface. Tantalum coating on titanium (TaTi) has exerted better tissue integration properties than TiI, but its biological performance under diabetic conditions remains elusive. To investigate whether TaTi may ameliorate diabetes-induced implant destabilization and the underlying mechanisms, primary rabbit osteoblasts cultured on 3-dimensional printed TiI and TaTi were exposed to normal serum (NS), diabetic serum (DS), DS+NAC (a potent ROS inhibitor), and DS+SB203580 (a specific p38 MAPK inhibitor). An in vivo study was performed on diabetic sheep implanted with TiI or TaTi. Diabetes induced mitochondrial-derived ROS overproduction and caused cellular dysfunction and apoptosis, together with the activation of p38 MAPK in osteoblasts on TiI surface. Importantly, TaTi significantly attenuated ROS production and p38 MAPK phosphorylation and exerted more osseointegrative cell behavior than TiI, as shown by improved osteoblast adhesion, increased cell proliferation and differentiation and decreased apoptosis. These results were confirmed in vivo by the enhanced bone healing efficacy of TaTi. Moreover, treatment with NAC or SB203580 on TiI not only inhibited the activation of p38 MAPK but also improved cell function and alleviated apoptotic injury, whereas TaTi combined with NAC or SB203580 failed to further improve osteoblast functional recovery compared with TaTi alone. These results demonstrated that the tantalum coating markedly improved diabetes-induced impaired osteogenesis of TiI, which may be attributed to the suppression of the ROS-mediated p38 MAPK pathway.