EMAIL THIS PAGE TO A FRIEND
Naunyn-Schmiedeberg's archives of pharmacology
CB
PMID 27659492
Abstract
We have shown previously that the cannabinoid receptor agonist CP55940 microinjected into the paraventricular nucleus of the hypothalamus (PVN) of urethane-anaesthetized rats induces depressor and pressor cardiovascular effects in the absence and presence of the CB
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3262
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
C5H9NO4