EMAIL THIS PAGE TO A FRIEND
Plant cell reports
Interaction between Cucumber mosaic virus 2b protein and plant catalase induces a specific necrosis in association with proteasome activity.
PMID 27659495
Abstract
Cucumber mosaic virus (CMV) can induce a specific necrosis on Arabidopsis through the interaction between the CMV 2b protein and host catalase, in which the ubiquitin-proteasome pathway may be involved. We previously reported that the CMV 2b protein, the viral RNA silencing suppressor, interacted with the H