EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Endogenous TRPV1 stimulation leads to the activation of the inositol phospholipid pathway necessary for sustained Ca
PMID 27663071
Abstract
Sensory neuron subpopulations as well as breast and prostate cancer cells express functional transient receptor potential vanilloid type 1 (TRPV1) ion channels; however little is known how TRPV1 activation leads to biological responses. Agonist-induced activation of TRPV1 resulted in specific spatiotemporal patterns of cytoplasmic Ca