EMAIL THIS PAGE TO A FRIEND
Ceskoslovenska fysiologie
[Pharmacokinetic parameters in old rats as a possible cause of side effects of drugs in old age].
PMID 2766379
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0383
Sulfameter
C11H12N4O3S
46834
Sulfameter, VETRANAL, analytical standard
C11H12N4O3S