EMAIL THIS PAGE TO A FRIEND
Journal of the science of food and agriculture
Characterization of peptides from common bean protein isolates and their potential to inhibit markers of type-2 diabetes, hypertension and oxidative stress.
PMID 27664971
Abstract
Diabetes and hypertension are diseases affecting a high proportion of the world population; the use of food-based products such as common bean peptides may contribute to reduce the risk of complications associated to chronic diseases. The aim was to produce and characterize peptides from common bean protein isolates and evaluate their potential to inhibit markers of type-2 diabetes, hypertension and oxidative stress. Mexican black and Brazilian Carioca bean isolated proteins were characterized after pepsin/pancreatin digestion. Also, four synthesized pure peptides, originally found in these beans, were evaluated. Bean protein digests and pure peptides exerted dipeptidyl peptidase-IV (DPP-IV) inhibition (IC Peptides from common bean have antidiabetic and antihypertensive potential regardless of their antioxidant capacity. © 2016 Society of Chemical Industry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9759
Ile-Pro-Ile, ≥97% (HPLC)
C17H31N3O4