EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
5,5-Dithiobis(2-nitrobenzoic acid) pyrene derivative-carbon nanotube electrodes for NADH electrooxidation and oriented immobilization of multicopper oxidases for the development of glucose/O
PMID 27665518
Abstract
We report the functionalization of multi-walled carbon nanotubes (MWCNTs) electrodes by a bifunctional nitroaromatic molecule accomplished via π-π interactions of a pyrene derivative. DTNB (5,5'-dithiobis(2-nitrobenzoic acid)) has the particularity to possess both electroactivable nitro groups and negatively charged carboxylic groups. The integration of the DTNB-modified MWCNTs was evaluated for different bioelectrocatalytic systems. The immobilized DTNB-based electrodes showed electrocatalytic activity toward the oxidation of the reduced form of nicotinamide adenine dinucleotide (NADH) with low overpotential of -0.09V vs Ag/AgCl at neutral pH. Glucose dehydrogenase was successfully immobilized at the surface of DTNB-based electrodes and, in the presence of NAD
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
401633
1-Pyrenemethylamine hydrochloride, 95%
C17H13N · HCl