EMAIL THIS PAGE TO A FRIEND
Biologicals : journal of the International Association of Biological Standardization
Development and application of HPLC-RI and HPLC-MS/MS based methods for quantification of residual deoxycholate levels in pneumococcal polysaccharides.
PMID 27666435
Abstract
The analysis of residual sodium deoxycholate (DOC); a detergent of biological origin used in manufacturing of polysaccharide vaccines is challenging due to complex sample matrices and the lack of suitable methods. Here we report, rapid and sensitive high-performance liquid chromatography-refractive index (HPLC-RI) and tandem mass spectrometry (HPLC-MS/MS) methods for estimation of residual DOC in pneumococcal polysaccharides. For HPLC-RI method, separation was achieved using Luna C18 column and mobile phase compositions of acetonitrile: methanol: 20 mM sodium acetate (60:05:35% v/v). For HPLC-MS/MS method, separation was achieved using a Hypersil BDS C18 column with gradient elution of methanol and water (0.1% formic acid). MS/MS method showed linearity (r
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6875
21-Hydroxyprogesterone, ≥97% (HPLC)
C21H30O3